| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200043091.mm.1 |
NM_001131021.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043091.mm.1 |
NM_177657.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043091.mm.1 |
TC0200005031.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200043089.mm.1 |
NM_001131021.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043089.mm.1 |
NM_177657.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043089.mm.1 |
TC0200005031.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200043088.mm.1 |
NM_001131021.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043088.mm.1 |
NM_177657.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043088.mm.1 |
TC0200005031.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200043090.mm.1 |
NM_001131021.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043090.mm.1 |
NM_177657.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200043090.mm.1 |
TC0200005031.mm.1 |
MTA TC |