Assay Details
Gene Symbol: Ano5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177694.6 | NP_808362.2 | 15 - 16 | 1808 | 105 | |
NR_073508.1 | 15 - 16 | 1801 | 105 | |||
GenBank mRNA | AB125740.1 | - | 15 - 16 | 1808 | 105 | |
AB206762.1 | - | 15 - 16 | 1808 | 105 | ||
AK034197.1 | - | 15 - 16 | 1790 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
