| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700008240.mm.1 |
TC0700000912.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008240.mm.1 |
NM_001271879.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008240.mm.1 |
NM_177694.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008240.mm.1 |
NR_073508.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008241.mm.1 |
TC0700000912.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008241.mm.1 |
NM_001271879.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008241.mm.1 |
NM_177694.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008241.mm.1 |
NR_073508.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008242.mm.1 |
TC0700000912.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700008242.mm.1 |
NM_001271879.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008242.mm.1 |
NM_177694.6 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700008242.mm.1 |
NR_073508.1 |
Refseq |