Assay Details
| Gene Symbol: Gdpd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177696.3 | NP_808364.2 | 1 - 2 | 80 | 84 | |
| GenBank mRNA | AK029527.1 | - | 1 - 2 | 83 | 84 | |
| AK161500.1 | - | 1 - 2 | 47 | 84 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029527.1, AK161500.1, NM_177696.3 transcripts.
Set Membership: |
|

Gdpd4
Genomic Map