Assay Details
| Gene Symbol: Vwa3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177697.3 | NP_808365.2 | 22 - 23 | 2678 | 103 | |
| GenBank mRNA | AK086942.1 | - | 21 - 22 | 2276 | 103 | |
| AK136898.1 | - | 21 - 22 | 2277 | 103 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Vwa3a
Genomic Map