Assay Details
| Gene Symbol: Ttll13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177765.3 | NP_808433.2 | 7 - 8 | 1145 | 70 | |
| GenBank mRNA | AK029565.1 | - | 7 - 8 | 1144 | 70 | |
| AK161519.1 | - | 7 - 8 | 968 | 70 | ||
| AM690757.1 | - | 6 - 7 | 846 | 70 | ||
| BC145749.1 | - | 6 - 7 | 753 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ttll13
Genomic Map