Assay Details
Gene Symbol: Ttll13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177765.3 | NP_808433.2 | 8 - 9 | 1234 | 101 | |
GenBank mRNA | AK029565.1 | - | 8 - 9 | 1233 | 101 | |
AK161519.1 | - | 8 - 9 | 1057 | 101 | ||
AM690757.1 | - | 7 - 8 | 935 | 101 | ||
BC145749.1 | - | 7 - 8 | 842 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
