Assay Details
Gene Symbol: Gm11961 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027798.2 | 2 - 3 | 599 | 63 | ||
GenBank mRNA | AK044388.1 | - | 2 - 3 | 601 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | NR_027797.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | NR_027798.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | AK044388.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | CN455772.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | CO427532.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | NR_027797.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | NR_027798.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | AK044388.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | CN455772.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P453995 | CO427532.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 132258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624226 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455140 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 423176 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 661134 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 160920 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 149547 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | NR_027797.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | NR_027798.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | AK044388.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | CN455772.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | CO427532.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | NR_027797.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | NR_027798.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | AK044388.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | CN455772.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P453995 | CO427532.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | NR_027797.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | NR_027798.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | AK044388.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | CN455772.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | CO427532.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | NR_027797.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | NR_027798.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | AK044388.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | CN455772.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P453995 | CO427532.1 | mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558661 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558621 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558619 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09009071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558682 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558640 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08995765 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09589543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09170051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558672 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09157221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558644 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558625 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558664 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558642 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558641 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558646 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558673 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558647 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558616 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558658 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09170052 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558623 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09514234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558624 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09647947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09009080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09392917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558671 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P09339418 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558639 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27558666 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10383886 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383885 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10383884 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10383883 | - | - |
Affymetrix Mouse Clariom S Array | PSR1100020568.mm.1 | NR_027798.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020568.mm.1 | NR_027798.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020568.mm.1 | TC1100002118.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1100020565.mm.1 | NR_027798.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020565.mm.1 | NR_027798.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1100020565.mm.1 | TC1100002118.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 730330 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243257 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243257 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243257 | - | - |
