Assay Details
Gene Symbol: Ubash3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177823.4 | NP_808491.2 | 10 - 11 | 1426 | 107 | |
GenBank mRNA | AK040572.1 | - | 10 - 11 | 1387 | 107 | |
AK041690.1 | - | 10 - 11 | 1393 | 107 | ||
AK049106.1 | - | 9 - 10 | 1326 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
