Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100027441.mm.1 |
NM_177839.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027441.mm.1 |
NM_177839.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027441.mm.1 |
TC0100003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027442.mm.1 |
NM_177839.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027442.mm.1 |
NM_177839.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027442.mm.1 |
TC0100003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027445.mm.1 |
NM_177839.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027445.mm.1 |
NM_177839.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027445.mm.1 |
TC0100003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027443.mm.1 |
NM_177839.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027443.mm.1 |
NM_177839.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027443.mm.1 |
TC0100003350.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100027444.mm.1 |
NM_177839.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027444.mm.1 |
NM_177839.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100027444.mm.1 |
TC0100003350.mm.1 |
MTA TC |