Assay Details
Gene Symbol: Tnn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177839.3 | NP_808507.2 | 8 - 9 | 1977 | 75 | |
GenBank mRNA | AF455756.1 | - | 8 - 9 | 1944 | 75 | |
AJ580920.1 | - | 7 - 8 | 1896 | 75 | ||
AK048033.1 | - | 1 - 2 | 92 | 75 | ||
BC138335.1 | - | 8 - 9 | 1938 | 8861355 | 75 | |
BC138336.1 | - | 8 - 9 | 1938 | 8861356 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
