Assay Details
| Gene Symbol: Spata21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177867.3 | NP_808535.1 | 3 - 4 | 485 | 61 | |
| GenBank mRNA | AK077155.1 | - | 3 - 4 | 486 | 61 | |
| BC112427.1 | - | 2 - 3 | 194 | 40062127 | 61 | |
| BC113132.1 | - | 2 - 3 | 194 | 40062125 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955188 | NM_177867.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1955188 | AK077155.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115310 | BC112427.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115310 | BC113132.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115310 | NM_177867.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2115310 | AK077155.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 405184 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 151421 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861954 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 268107 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637577 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 624589 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373768 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11209 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 488527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 81182 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 33571 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120952 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415420 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 702741 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869857 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759257 | AK050955.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759257 | AK077394.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759257 | AK161040.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 759257 | NM_028248.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65786 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 144976 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108667 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P155255 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108031 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115310 | BC112427.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115310 | BC113132.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115310 | NM_177867.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2115310 | AK077155.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955188 | NM_177867.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1955188 | AK077155.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23362056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23362063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23362059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23362060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23362054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23362064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23362057 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01006844 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01006857 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P155255 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108031 | - | - |
| Whole Mouse Genome G4122A | A_51_P210059 | BC112427.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210059 | BC113132.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210059 | NM_177867.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P210059 | AK077155.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131736 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108031 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01006844 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P155255 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03224122 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03224123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362018 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362167 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362236 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362072 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362248 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362076 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362231 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362030 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362240 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362171 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362058 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362029 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362187 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362266 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362245 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362031 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362028 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362264 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362134 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362068 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362222 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362032 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23362180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362165 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362189 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03961290 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362180 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362178 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04066020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04066021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362052 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03604924 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03933193 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362170 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03986845 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362179 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03811824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03441356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03623431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04066026 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03894226 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03521503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362073 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03551166 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362183 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362058 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362190 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362072 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03441352 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03441357 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362056 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03623426 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04063969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362080 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03623429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03753756 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03986844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03523906 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23362060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03858373 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03835175 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10509918 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509911 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509917 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509922 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509919 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509916 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509925 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509924 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509923 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509913 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509912 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509915 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509926 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509914 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509920 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10509921 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10509910 | - | - |
| Mouse Genome 430 2.0 Array | 1455718_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1455718_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329972_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 329972_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0400014705.mm.1 | NM_177867.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014705.mm.1 | NM_177867.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014705.mm.1 | TC0400001777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014706.mm.1 | NM_177867.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014706.mm.1 | NM_177867.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014706.mm.1 | TC0400001777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014707.mm.1 | NM_177867.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014707.mm.1 | NM_177867.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014707.mm.1 | TC0400001777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014703.mm.1 | NM_177867.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014703.mm.1 | NM_177867.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014703.mm.1 | TC0400001777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014702.mm.1 | NM_177867.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014702.mm.1 | NM_177867.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014702.mm.1 | TC0400001777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014704.mm.1 | NM_177867.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014704.mm.1 | NM_177867.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014704.mm.1 | TC0400001777.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0400014708.mm.1 | NM_177867.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014708.mm.1 | NM_177867.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0400014708.mm.1 | TC0400001777.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455718_PM_at | - | - |
| Mouse Expression Set 430 | 1455718_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6840091 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6840091 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006840091 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2626950 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2626950 | - | - |
| mouse-6 v1.1 (Illumina) | scl24755.16.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6840091 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl24755.16.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6840091 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2626950 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2626950 | - | - |

Spata21
Genomic Map