Assay Details
| Gene Symbol: Rnf212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089585.1 | - | 5 - 6 | 617 | 80 | |
| BC109180.2 | - | 5 - 6 | 576 | 80 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2116350 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77227 | AK089585.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77227 | BC109180.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77227 | AK089585.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P77227 | BC109180.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P156030 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77227 | AK089585.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77227 | BC109180.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77227 | AK089585.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P77227 | BC109180.2 | mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973969 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04620186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04176461 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973842 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973978 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973857 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04739040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973988 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973959 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04940186 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04176444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973848 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973995 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973979 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04704966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04620191 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04375020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973939 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973984 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973945 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04176452 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973965 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04226824 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04819989 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04850588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973849 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973834 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973966 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04176436 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973846 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253769 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04940196 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973951 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973975 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04176439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04850587 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973831 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973993 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973833 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973973 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973933 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973841 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973967 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P23973983 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04253804 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10532249 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532250 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532257 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532253 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532260 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532259 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532254 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532258 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10532256 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10532248 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 399150 | - | - |

Rnf212
Genomic Map