Assay Details
| Gene Symbol: Gm9801 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086712.1 | - | 2543 | 84 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P305721 | AK086712.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P305721 | AK086712.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P111843 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168031 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168000 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168029 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168010 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168006 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168033 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167985 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168020 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168025 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167999 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168022 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167998 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168035 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168017 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25167992 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25168026 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P111843 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215155 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215197 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215175 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215280 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215167 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215253 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215127 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215210 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01529043 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215215 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215230 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06215271 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101157 | - | - |
| Whole Mouse Genome G4122A | A_51_P305721 | AK086712.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P305721 | AK086712.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127656 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117879 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01529043 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06215253 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P111843 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06215215 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101157 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168022 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167969 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168006 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167998 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167971 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167977 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168042 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167976 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05919719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168003 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167984 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168040 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168019 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167993 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168025 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168029 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167974 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167970 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168016 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167983 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167967 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168020 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06014073 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25167988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168037 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25168013 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 301736 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5910603 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5910603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1236551 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236551 | - | - |
| mouse-6 v1.1 (Illumina) | scl31262.22.1_71-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910603 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl31262.22.1_71-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910603 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1236551 | - | - |

Gm9801
Genomic Map