Assay Details
| Gene Symbol: Gmppb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177910.3 | NP_808578.1 | 8 - 9 | 1180 | 82 | |
| GenBank mRNA | AK088295.1 | - | 8 - 9 | 1162 | 82 | |
| AK148125.1 | - | 1180 | 82 | |||
| AK158470.1 | - | 8 - 9 | 1180 | 82 | ||
| AK165816.1 | - | 7 - 8 | 1265 | 82 | ||
| BC061207.1 | - | 9 - 10 | 1054 | 30249330 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812599 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812600 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812602 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812601 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812604 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | AK158470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | AK165816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | BC061207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | NM_177910.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | AK088295.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | AK158470.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | AK165816.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | BC061207.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | NM_177910.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P386810 | AK088295.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675336 | AK047478.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675336 | AK081604.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675336 | AK087509.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675336 | BC017649.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675336 | BC051682.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 675336 | NM_001015681.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 191343 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P151500 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | AK158470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | AK165816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | BC061207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | NM_177910.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | AK088295.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | AK158470.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | AK165816.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | BC061207.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | NM_177910.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P386810 | AK088295.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812623 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812599 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812603 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812612 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812613 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812615 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812610 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812604 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812598 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812620 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812616 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26812618 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P151500 | - | - |
| Whole Mouse Genome G4122A | A_51_P386810 | AK158470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | AK165816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | BC061207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | NM_177910.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | AK088295.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | AK158470.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | AK165816.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | BC061207.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | NM_177910.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P386810 | AK088295.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115111 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P151500 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07452570 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07452522 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812604 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812603 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812602 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812600 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812599 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26812601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812654 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812605 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812656 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812628 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812622 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812638 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812615 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812636 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812616 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812643 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812645 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812610 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812632 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812598 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26812623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812625 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812623 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812607 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812614 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812603 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07452626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812599 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812605 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26812618 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10588838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10588845 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1439030_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10588836 | - | - |
| Mouse Genome 430 2.0 Array | 1439030_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1439030_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 331026_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 331026_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900011157.mm.1 | NM_177910.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011157.mm.1 | NM_177910.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011157.mm.1 | TC0900001391.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900011155.mm.1 | NM_177910.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011155.mm.1 | NM_177910.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011155.mm.1 | TC0900001391.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900011154.mm.1 | NM_177910.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011154.mm.1 | NM_177910.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011154.mm.1 | TC0900001391.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900011158.mm.1 | NM_177910.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011158.mm.1 | NM_177910.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011158.mm.1 | TC0900001391.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1439030_PM_at | - | - |
| Mouse Expression Set 430 | 1439030_at | - | - |
| Murine Genome U74v2 Set | 104351_at | - | - |
| Murine Genome U74v2 Set | 104351_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439030_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1439030_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439030_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1439030_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 623205 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4610538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4610538 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004610538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2744910 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2744910 | - | - |
| mouse-6 v1.1 (Illumina) | scl36480.8.1_52-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4610538 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36480.8.1_52-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4610538 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2744909 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2744909 | - | - |

Gmppb
Genomic Map