Assay Details
| Gene Symbol: Stard9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029170.1 | - | 2260 | 126 | ||
| AK029732.1 | - | 10 - 11 | 3609 | 126 | ||
| AK043337.1 | - | 1 - 2 | 447 | 126 | ||
| AK122489.1 | - | 10 - 11 | 4309 | 126 | ||
| BC032885.1 | - | 8 - 9 | 1019 | 126 | ||
| BC034905.1 | - | 1 - 2 | 355 | 126 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044533 | AK122489.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2098086 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P137439 | AB001425.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2238720 | BC032885.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2238720 | AK029732.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2238720 | AK029170.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2238720 | AK122489.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451970 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 61737 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P128563 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178064 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P108017 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P158835 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P154287 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2238720 | BC032885.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2238720 | AK029732.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2238720 | AK029170.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2238720 | AK122489.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P137439 | AB001425.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044533 | AK122489.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P158835 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P128563 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P108017 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04668776 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P154287 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178064 | - | - |
| Whole Mouse Genome G4122A | A_51_P270583 | AK087617.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P233687 | BC034905.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P233687 | BC032885.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P233687 | AK029732.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P233687 | AK043337.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P233687 | AK049942.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P376501 | AK029170.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410312 | AK138464.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P410312 | AB001425.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105059 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P128131 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137439 | AB001425.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P158835 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P154287 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P108017 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178064 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P128563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01400905 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576796 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576726 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576713 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576576 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01652955 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576498 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576553 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576732 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02114595 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576733 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576784 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576617 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576735 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576513 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576550 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576775 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576790 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576624 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576583 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01532744 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576766 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576665 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576563 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576676 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01694754 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576537 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01454864 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576662 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576700 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576557 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576608 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01532736 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724890 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576782 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01928623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724868 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576527 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02004661 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02154508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576508 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01647188 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576494 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576760 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01625851 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01759535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02220634 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02185325 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576538 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01736081 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576801 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576534 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01466053 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02246129 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01532742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576568 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576545 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576753 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01738641 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576579 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01652957 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576683 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01532738 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576798 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576603 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576745 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576585 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01532750 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576602 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576718 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02114599 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01466051 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01454862 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576647 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576674 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576633 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01463521 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576589 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576639 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724818 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01551779 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576670 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576697 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576792 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576524 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576565 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576566 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576742 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576780 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576708 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576648 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576506 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576573 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576772 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576570 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01625843 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576702 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576793 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01451227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724867 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576668 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576529 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01153202 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576799 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724789 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576588 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576764 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724847 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576619 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576728 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01400909 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576706 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724850 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576593 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576640 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576757 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01681657 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02004660 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02206836 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576658 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576577 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576759 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01151852 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576730 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576546 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724898 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576516 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576611 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01400907 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576677 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01532746 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576710 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576675 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576502 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01543485 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576560 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01451230 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724876 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576699 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01543483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576649 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576561 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576558 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01463520 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01625844 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576554 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01445075 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724854 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01445078 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576578 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576582 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01532734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01463518 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724828 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576523 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21576645 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01724861 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01652960 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10475232 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475243 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475238 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475242 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475233 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475234 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475240 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475241 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475239 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475235 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475230 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475237 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475244 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10475236 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1459437_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436324_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10475229 | - | - |
| Mouse Genome 430 2.0 Array | 1436324_at | - | - |
| Mouse Genome 430 2.0 Array | 1436324_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459437_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1459437_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 668880_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 668880_PM_at | - | - |
| Mouse Expression Set 430 | 1436324_PM_at | - | - |
| Mouse Expression Set 430 | 1459437_PM_at | - | - |
| Mouse Expression Set 430 | 1436324_at | - | - |
| Mouse Expression Set 430 | 1459437_at | - | - |
| Murine Genome U74v2 Set | 140759_PM_at | - | - |
| Murine Genome U74v2 Set | 140759_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436324_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459437_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1459437_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1436324_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 102970037 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102970037 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102970037 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1254226 | - | - |

Stard9
Genomic Map