Assay Details
| Gene Symbol: Pde4dip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039376.2 | NP_001034465.2 | 13 - 14 | 1752 | 75 | |
| NM_001289701.1 | NP_001276630.1 | 13 - 14 | 1752 | 75 | ||
| NM_001289702.1 | NP_001276631.1 | 13 - 14 | 1752 | 75 | ||
| NM_178080.4 | NP_835181.2 | 6 - 7 | 2260 | 75 | ||
| NR_110360.1 | 6 - 7 | 2774 | 75 | |||
| GenBank mRNA | AK028810.1 | - | 6 - 7 | 2227 | 75 | |
| AK122286.1 | - | 6 - 7 | 2255 | 75 | ||
| AK141917.1 | - | 13 - 14 | 1878 | 75 | ||
| BC025653.1 | - | 6 - 7 | 2186 | 75 | ||
| BC028274.1 | - | 13 - 14 | 1762 | 75 | ||
| BC034266.1 | - | 6 - 7 | 925 | 75 | ||
| BC050783.1 | - | 6 - 7 | 2214 | 75 | ||
| BC057350.1 | - | 6 - 7 | 2691 | 75 | ||
More Information
Set Membership: |
|

Pde4dip
Genomic Map