Assay Details
| Gene Symbol: Tcfl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178254.3 | NP_839985.2 | 4 - 5 | 1309 | 61 | |
| GenBank mRNA | AY234363.1 | - | 4 - 5 | 1103 | 61 | |
| BC108399.1 | - | 4 - 5 | 1309 | 6394526 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905265 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905269 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905262 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 415379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 330833 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 88305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875241 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK005012.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK015410.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK154375.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK157844.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK170574.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK171049.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK171566.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK172231.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK180287.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | AK206038.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | L31398.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654365 | NM_001039520.1 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630140 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579288 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 417632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 169872 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139766 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116573 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905262 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905251 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905259 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905260 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905282 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905284 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905255 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905257 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905265 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905275 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905247 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905256 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905285 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905271 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905278 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905253 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905270 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905274 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21905263 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116573 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00573612 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139766 | - | - |
| Whole Mouse Genome G4122A | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299713 | BC108399.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299713 | NM_178254.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P299713 | AY234363.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109994 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119517 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116573 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139766 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01399230 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905263 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905267 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905269 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905262 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21905265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905181 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905262 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905246 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905213 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905226 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905115 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905117 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905112 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905194 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905108 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905159 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905274 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905219 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905284 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905106 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905083 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905172 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905102 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905242 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21905249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905271 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905284 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905231 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905275 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02218040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905234 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905245 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905224 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01452115 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905265 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01741544 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905215 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905228 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01471519 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01254214 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905274 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01741536 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905221 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905253 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905277 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905247 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905249 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905267 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905222 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905279 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905251 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905241 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905272 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905255 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905219 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905273 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01468921 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01360936 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905244 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01339042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905256 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21905227 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10490509 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490507 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490508 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490505 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490506 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490503 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490504 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1456515_PM_s_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10479423 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10490502 | - | - |
| Mouse Genome 430 2.0 Array | 1456515_s_at | - | - |
| Mouse Genome 430 2.0 Array | 1456515_PM_s_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277353_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 277353_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200046104.mm.1 | NM_178254.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200046104.mm.1 | NM_178254.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200046104.mm.1 | TC0200005414.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1456515_s_at | - | - |
| Mouse Expression Set 430 | 1456515_PM_s_at | - | - |
| Murine Genome U74v2 Set | 165998_PM_at | - | - |
| Murine Genome U74v2 Set | 165998_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456515_s_at | - | - |
| Mouse Genome 430A 2.0 Array | 1456515_PM_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456515_s_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1456515_PM_s_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 725980 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6180286 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6180286 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006180286 | - | - |
| mouse-6 v1.1 (Illumina) | scl18225.6.1_262-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6180286 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl18225.6.1_262-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6180286 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2972794 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733257 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2972794 | - | - |

Tcfl5
Genomic Map