Assay Details
| Gene Symbol: Gulo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178747.3 | NP_848862.1 | 1 - 2 | 167 | 67 | |
| GenBank mRNA | AK077740.1 | - | 1 - 2 | 168 | 67 | |
| AK167460.1 | - | 1 - 2 | 156 | 67 | ||
| BC019856.1 | - | 1 - 2 | 114 | 5123684 | 67 | |
| BC028822.1 | - | 1 - 2 | 123 | 5097681 | 67 | |
| BC028828.1 | - | 1 - 2 | 126 | 5101228 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK077740.1, AK167460.1, BC019856.1, BC028822.1, BC028828.1, NM_178747.3 transcripts.
Set Membership: |
|

Gulo
Genomic Map