Assay Details
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178765.3 | NP_848880.2 | 1 - 2 | 373 | 143 | |
| GenBank mRNA | AB232449.1 | - | 1 - 2 | 373 | 143 | |
| AK030482.1 | - | 1 - 2 | 105 | 143 | ||
| AK030720.1 | - | 1 - 2 | 105 | 143 | ||
| AK163432.1 | - | 1 - 2 | 138 | 143 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB232449.1, AK030482.1, AK030720.1, AK163432.1, NM_178765.3 transcripts.
Set Membership: |
|

Sybu
Genomic Map