Assay Details
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285840.1 | NP_001272769.1 | 1 - 2 | 362 | 69 | |
| NM_176998.4 | NP_795972.2 | 2 - 3 | 563 | 69 | ||
| NM_178765.3 | NP_848880.2 | 2 - 3 | 460 | 69 | ||
| GenBank mRNA | AB232447.1 | - | 2 - 3 | 563 | 69 | |
| AB232449.1 | - | 2 - 3 | 460 | 69 | ||
| AK030482.1 | - | 2 - 3 | 192 | 69 | ||
| AK030720.1 | - | 2 - 3 | 192 | 69 | ||
| AK163432.1 | - | 2 - 3 | 225 | 69 | ||
| AK220268.1 | - | 1 - 2 | 362 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB232449.1, AK030482.1, AK030720.1, AK163432.1, NM_001285840.1, NM_178765.3 transcripts.
Set Membership: |
|

Sybu
Genomic Map