Assay Details
| Gene Symbol: Sbf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177324.2 | NP_796298.2 | 7 - 8 | 906 | 76 | |
| GenBank mRNA | AK046622.1 | - | 7 - 8 | 894 | 76 | |
| AK048952.1 | - | 7 - 8 | 883 | 76 | ||
| AK080952.1 | - | 7 - 8 | 907 | 76 | ||
| AK080963.1 | - | 7 - 8 | 901 | 76 | ||
| AK087396.1 | - | 7 - 8 | 901 | 76 | ||
| AK173257.1 | - | 7 - 8 | 865 | 76 | ||
| BC048706.1 | - | 7 - 8 | 891 | 76 | ||
| BC067204.1 | - | 8 - 9 | 944 | 76 | ||
More Information
Set Membership: |
|

Sbf2
Genomic Map