Assay Details
| Gene Symbol: Slc7a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178798.3 | NP_848913.1 | 8 - 9 | 1388 | 73 | |
| GenBank mRNA | AK030762.1 | - | 9 - 10 | 1190 | 73 | |
| AK046548.1 | - | 7 - 8 | 2055 | 73 | ||
| AK049478.1 | - | 8 - 9 | 1388 | 73 | ||
| AK053899.1 | - | 8 - 9 | 1397 | 73 | ||
| AK088592.1 | - | 8 - 9 | 1305 | 73 | ||
| AK129099.1 | - | 5 - 6 | 2784 | 73 | ||
| AK157127.1 | - | 8 - 9 | 1527 | 73 | ||
| AK157379.1 | - | 7 - 8 | 2057 | 73 | ||
| AK164666.1 | - | 8 - 9 | 1389 | 73 | ||
| AK165629.1 | - | 8 - 9 | 1405 | 73 | ||
| AK165979.1 | - | 8 - 9 | 1407 | 73 | ||
| BC038404.1 | - | 7 - 8 | 1323 | 4949962 | 73 | |
| BC042898.1 | - | 5 - 6 | 1920 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a6
Genomic Map