Assay Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178911.4 | NP_849242.1 | 1 - 2 | 133 | 84 | |
| GenBank mRNA | AK045459.1 | - | 1 - 2 | 92 | 84 | |
| AK079549.1 | - | 1 - 2 | 92 | 84 | ||
| AK079878.1 | - | 1 - 2 | 135 | 84 | ||
| AK170362.1 | - | 1 - 2 | 90 | 84 | ||
| BC058565.1 | - | 1 - 2 | 83 | 6442038 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK045459.1, AK079549.1, AK079878.1, AK170362.1, BC058565.1, NM_178911.4 transcripts.
Set Membership: |
|

Pld4
Genomic Map