Assay Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178911.4 | NP_849242.1 | 4 - 5 | 564 | 87 | |
| GenBank mRNA | AK045459.1 | - | 4 - 5 | 523 | 87 | |
| AK079549.1 | - | 4 - 5 | 523 | 87 | ||
| AK079878.1 | - | 4 - 5 | 566 | 87 | ||
| AK151551.1 | - | 4 - 5 | 798 | 87 | ||
| AK152969.1 | - | 3 - 4 | 411 | 87 | ||
| AK170362.1 | - | 4 - 5 | 521 | 87 | ||
| BC058565.1 | - | 4 - 5 | 514 | 6442038 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pld4
Genomic Map