Assay Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178911.4 | NP_849242.1 | 5 - 6 | 683 | 74 | |
| GenBank mRNA | AK045459.1 | - | 5 - 6 | 642 | 74 | |
| AK079549.1 | - | 5 - 6 | 642 | 74 | ||
| AK079878.1 | - | 5 - 6 | 685 | 74 | ||
| AK151551.1 | - | 5 - 6 | 917 | 74 | ||
| AK152969.1 | - | 4 - 5 | 530 | 74 | ||
| AK170362.1 | - | 5 - 6 | 640 | 74 | ||
| BC058565.1 | - | 5 - 6 | 633 | 6442038 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pld4
Genomic Map