Assay Details
| Gene Symbol: Pld4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178911.4 | NP_849242.1 | 8 - 9 | 1151 | 63 | |
| GenBank mRNA | AK045459.1 | - | 8 - 9 | 1110 | 63 | |
| AK079549.1 | - | 8 - 9 | 1110 | 63 | ||
| AK079878.1 | - | 8 - 9 | 1153 | 63 | ||
| AK151551.1 | - | 8 - 9 | 1385 | 63 | ||
| AK152969.1 | - | 7 - 8 | 999 | 64 | ||
| AK170362.1 | - | 8 - 9 | 1108 | 63 | ||
| BC058565.1 | - | 8 - 9 | 1101 | 6442038 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pld4
Genomic Map