Assay Details
| Gene Symbol: Lmf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178919.4 | NP_849250.1 | 1 - 2 | 299 | 76 | |
| GenBank mRNA | AK042427.1 | - | 1 - 2 | 258 | 76 | |
| AK084606.1 | - | 1 - 2 | 125 | 76 | ||
| AK085435.1 | - | 1 - 2 | 250 | 76 | ||
| AK149916.1 | - | 1 - 2 | 251 | 76 | ||
| AK149934.1 | - | 1 - 2 | 300 | 76 | ||
| AK158371.1 | - | 1 - 2 | 273 | 76 | ||
| AK171488.1 | - | 1 - 2 | 280 | 76 | ||
| AK184485.1 | - | 1 - 2 | 42 | 76 | ||
| AK185616.1 | - | 1 - 2 | 42 | 76 | ||
| BC079532.1 | - | 1 - 2 | 195 | 6403184 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424718 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424714 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424716 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC079532.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC038039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC058973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | AV224384.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC026448.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC079532.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC038039.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC058973.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | AV224384.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P54238 | BC026448.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406246 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487381 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340921 | AK077052.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340921 | AK132066.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340921 | AY069976.2 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340921 | BC020286.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340921 | BC029632.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340921 | BC059282.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 340921 | NM_172705.2 | refseq_mrna_mouse_NM |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P167906 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC079532.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC038039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC058973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | AV224384.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC026448.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC079532.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC038039.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC058973.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | AV224384.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P54238 | BC026448.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424694 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424679 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424674 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424700 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424676 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424713 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424701 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424690 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424710 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30424680 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P167906 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03096350 | - | - |
| Whole Mouse Genome G4122A | A_52_P54238 | BC079532.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | BC038039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | BC058973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | AV224384.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | BC026448.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | BC079532.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | BC038039.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | BC058973.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | AV224384.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P54238 | BC026448.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131750 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P167906 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11870881 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11870895 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11870850 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11870893 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P11870894 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424716 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424714 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424717 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30424718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424710 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424686 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424712 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424663 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30424649 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424692 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424660 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424699 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11870877 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424705 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11870893 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11870884 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424713 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424663 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11870856 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11870885 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424712 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424710 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424686 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424675 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424706 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424670 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424661 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424690 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P11870864 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424674 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424717 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424667 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30424696 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10431535 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431543 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431533 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431530 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431537 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431542 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431538 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431536 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431541 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431529 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431531 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431534 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431544 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431532 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431540 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10431539 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1454619_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1433100_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10431528 | - | - |
| Mouse Genome 430 2.0 Array | 1454619_at | - | - |
| Mouse Genome 430 2.0 Array | 1454619_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105847_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 105847_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1500016478.mm.1 | NM_178919.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016478.mm.1 | NM_178919.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016478.mm.1 | TC1500002050.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500016481.mm.1 | NM_178919.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016481.mm.1 | NM_178919.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016481.mm.1 | TC1500002050.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500016483.mm.1 | NM_178919.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016483.mm.1 | NM_178919.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016483.mm.1 | TC1500002050.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500016479.mm.1 | NM_178919.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016479.mm.1 | NM_178919.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016479.mm.1 | TC1500002050.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1500016482.mm.1 | NM_178919.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016482.mm.1 | NM_178919.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1500016482.mm.1 | TC1500002050.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1454619_PM_at | - | - |
| Mouse Expression Set 430 | 1454619_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433100_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433100_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454619_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1454619_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 797611 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4670603 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004670603 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2736566 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2736566 | - | - |
| mouse-6 v1.1 (Illumina) | scl46854.10.1_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4670603 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl46854.10.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670603 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2736566 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2736566 | - | - |

Lmf2
Genomic Map