Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500002137.mm.1 |
NM_008056.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002137.mm.1 |
NM_008056.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002137.mm.1 |
TC1500000298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002138.mm.1 |
NM_001162494.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002138.mm.1 |
NM_001162494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002138.mm.1 |
NM_008056.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002138.mm.1 |
NM_008056.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002138.mm.1 |
TC1500000298.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1500002136.mm.1 |
NM_001162494.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002136.mm.1 |
NM_001162494.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002136.mm.1 |
NM_008056.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002136.mm.1 |
NM_008056.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500002136.mm.1 |
TC1500000298.mm.1 |
MTA TC |