Assay Details
| Gene Symbol: Smad1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008539.3 | NP_032565.2 | 1 - 2 | 220 | 151 | |
| GenBank mRNA | AK017583.1 | - | 1 - 2 | 181 | 151 | |
| AK034045.1 | - | 1 - 2 | 181 | 151 | ||
| AK054104.1 | - | 1 - 2 | 181 | 151 | ||
| AK084505.1 | - | 1 - 2 | 288 | 152 | ||
| AK087692.1 | - | 181 | 151 | |||
| AK158509.1 | - | 1 - 2 | 283 | 151 | ||
| BC058693.1 | - | 1 - 2 | 220 | 6811514 | 151 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK017583.1, AK034045.1, AK054104.1, AK084505.1, AK087692.1, BC058693.1, NM_008539.3 transcripts.
Set Membership: |
|

Smad1
Genomic Map