Assay Details
| Gene Symbol: Man1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008548.4 | NP_032574.1 | 7 - 8 | 1646 | 136 | |
| GenBank mRNA | AK031038.1 | - | 7 - 8 | 1647 | 136 | |
| AK042371.1 | - | 6 - 7 | 1487 | 136 | ||
| AK043932.1 | - | 7 - 8 | 1587 | 136 | ||
| AK154820.1 | - | 7 - 8 | 1588 | 136 | ||
| AK171841.1 | - | 7 - 8 | 1589 | 136 | ||
| BC015265.1 | - | 7 - 8 | 1547 | 4223319 | 136 | |
| U04299.1 | - | 6 - 7 | 1250 | 136 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Man1a
Genomic Map