Assay Details
Gene Symbol: Mas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008552.4 | NP_032578.2 | 3 - 4 | 331 | 74 | |
GenBank mRNA | AK015036.1 | - | 3 - 4 | 322 | 74 | |
AK030261.1 | - | 3 - 4 | 321 | 74 | ||
AK030265.1 | - | 3 - 4 | 332 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK015036.1, AK030261.1, AK030265.1, NM_008552.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
