Assay Details
Gene Symbol: Mas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008552.4 | NP_032578.2 | 8 - 9 | 908 | 58 | |
GenBank mRNA | AK015036.1 | - | 8 - 9 | 899 | 58 | |
AK030261.1 | - | 8 - 9 | 898 | 58 | ||
AK030265.1 | - | 8 - 9 | 909 | 58 | ||
BC120790.1 | - | 3 - 4 | 230 | 40129713 | 58 | |
BC125537.1 | - | 4 - 5 | 293 | 40130052 | 58 | |
BC144900.1 | - | 3 - 4 | 242 | 9053448 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK015036.1, AK030261.1, AK030265.1, BC120790.1, BC125537.1, BC144900.1, NM_008552.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
