Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_001166388.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_001166388.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_001166389.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_001166389.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_001166390.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_001166390.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_008975.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
NM_008975.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1500003664.mm.1 |
TC1500000524.mm.1 |
MTA TC |