Assay Details
| Gene Symbol: Tgm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009373.3 | NP_033399.1 | 1 - 2 | 97 | 100 | |
| GenBank mRNA | AF114266.1 | - | 1 - 2 | 62 | 100 | |
| AK052912.1 | - | 1 - 2 | 72 | 100 | ||
| AK054108.1 | - | 1 - 2 | 72 | 100 | ||
| AK080224.1 | - | 1 - 2 | 97 | 100 | ||
| AK080593.1 | - | 1 - 2 | 72 | 100 | ||
| AK086001.1 | - | 1 - 2 | 72 | 100 | ||
| AK089481.1 | - | 1 - 2 | 100 | 100 | ||
| AK143712.1 | - | 1 - 2 | 133 | 100 | ||
| AK151776.1 | - | 1 - 2 | 98 | 100 | ||
| AK152152.1 | - | 1 - 2 | 72 | 100 | ||
| AK152627.1 | - | 1 - 2 | 98 | 100 | ||
| AK159255.1 | - | 1 - 2 | 72 | 100 | ||
| AK166302.1 | - | 1 - 2 | 72 | 100 | ||
| AK168990.1 | - | 1 - 2 | 73 | 100 | ||
| AK169356.1 | - | 1 - 2 | 100 | 100 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF114266.1, AK052912.1, AK080224.1, AK080593.1, AK089481.1, AK143712.1, AK151776.1, AK152152.1, AK152627.1, AK159255.1, AK166302.1, AK168990.1, AK169356.1, NM_009373.3 transcripts.
Set Membership: |
|

Tgm2
Genomic Map