Assay Details
| Gene Symbol: Traf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009423.4 | NP_033449.2 | 4 - 5 | 568 | 88 | |
| GenBank mRNA | AF233449.1 | - | 4 - 5 | 467 | 88 | |
| AK035056.1 | - | 4 - 5 | 546 | 88 | ||
| AK040877.1 | - | 546 | 88 | |||
| AK046015.1 | - | 4 - 5 | 608 | 88 | ||
| AK053900.1 | - | 4 - 5 | 565 | 88 | ||
| AK184213.1 | - | 2 - 3 | 215 | 88 | ||
| BC020455.1 | - | 4 - 5 | 553 | 88 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Traf4
Genomic Map