Assay Details
Gene Symbol: Emp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001288628.1 | NP_001275557.1 | 1 - 2 | 169 | 83 | |
NM_010128.4 | NP_034258.1 | 1 - 2 | 169 | 83 | ||
GenBank mRNA | AK157366.1 | - | 1 - 2 | 87 | 83 | |
AK158369.1 | - | 1 - 2 | 108 | 83 | ||
AK159752.1 | - | 1 - 2 | 87 | 83 | ||
AK168806.1 | - | 1 - 2 | 61 | 83 | ||
AK171061.1 | - | 1 - 2 | 87 | 83 | ||
AK171675.1 | - | 1 - 2 | 87 | 83 | ||
AK172607.1 | - | 1 - 2 | 87 | 83 | ||
AK172705.1 | - | 1 - 2 | 87 | 83 | ||
BC034257.1 | - | 1 - 2 | 69 | 3157463 | 83 | |
BC046299.1 | - | 1 - 2 | 169 | 6486958 | 83 | |
BC138491.1 | - | 1 - 2 | 30 | 8861511 | 83 | |
U25633.1 | - | 1 - 2 | 53 | 83 | ||
X98403.1 | - | 1 - 2 | 146 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK157366.1, AK159752.1, AK168806.1, AK171061.1, AK171675.1, AK172607.1, AK172705.1, BC034257.1, BC046299.1, BC138491.1, NM_001288628.1, NM_010128.4, U25633.1, X98403.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
