Assay Details
Gene Symbol: Kcnab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289450.1 | NP_001276379.1 | 11 - 12 | 797 | 102 | |
NM_010597.4 | NP_034727.3 | 10 - 11 | 1213 | 102 | ||
GenBank mRNA | AF033003.1 | - | 10 - 11 | 1568 | 102 | |
AK004666.1 | - | 11 - 12 | 781 | 102 | ||
AK015412.1 | - | 12 - 13 | 1053 | 102 | ||
AK031362.1 | - | 10 - 11 | 1214 | 102 | ||
AK138467.1 | - | 10 - 11 | 1202 | 102 | ||
AK153200.1 | - | 11 - 12 | 799 | 102 | ||
BC014701.1 | - | 10 - 11 | 938 | 4482268 | 102 | |
X97281.1 | - | 10 - 11 | 1197 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
