Assay Details
Gene Symbol: Ncam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113208.1 | NP_001106679.1 | 10 - 11 | 1599 | 126 | |
NM_010954.4 | NP_035084.1 | 10 - 11 | 1599 | 126 | ||
GenBank mRNA | AF001286.1 | - | 10 - 11 | 1525 | 126 | |
AF001287.1 | - | 10 - 11 | 1525 | 126 | ||
AF016619.1 | - | 10 - 11 | 1481 | 126 | ||
BC119027.1 | - | 10 - 11 | 1457 | 8733776 | 126 | |
BC119029.1 | - | 10 - 11 | 1457 | 8733778 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
