Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164480.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164480.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164481.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164481.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164482.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164482.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164568.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_001164568.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_011379.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
NM_011379.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1900008721.mm.1 |
TC1900000977.mm.1 |
MTA TC |