Assay Details
| Gene Symbol: Ppp2ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019411.4 | NP_062284.1 | 3 - 4 | 710 | 59 | |
| GenBank mRNA | AF076192.1 | - | 3 - 4 | 680 | 59 | |
| AK076110.1 | - | 3 - 4 | 711 | 59 | ||
| AK172644.1 | - | 3 - 4 | 683 | 59 | ||
| BC003856.1 | - | 3 - 4 | 687 | 3484084 | 59 | |
| BC054458.1 | - | 3 - 4 | 670 | 6493866 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2ca
Genomic Map