Assay Details
| Gene Symbol: Plxdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026162.6 | NP_080438.2 | 8 - 9 | 1801 | 138 | |
| GenBank mRNA | AF378761.1 | - | 8 - 9 | 1000 | 138 | |
| AK004640.1 | - | 8 - 9 | 1783 | 138 | ||
| AK019482.2 | - | 8 - 9 | 1757 | 138 | ||
| AK035165.1 | - | 8 - 9 | 1803 | 138 | ||
| AK154921.1 | - | 8 - 9 | 1748 | 138 | ||
| BC057881.1 | - | 8 - 9 | 1061 | 5349869 | 138 | |
More Information
Set Membership: |
|

Plxdc2
Genomic Map