Assay Details
| Gene Symbol: Irak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028679.3 | NP_082955.2 | 4 - 5 | 552 | 121 | |
| GenBank mRNA | AJ440757.2 | - | 4 - 5 | 474 | 121 | |
| AK014783.1 | - | 4 - 5 | 603 | 121 | ||
| AK028436.1 | - | 465 | 121 | |||
| AK029057.1 | - | 4 - 5 | 553 | 121 | ||
| AK049210.1 | - | 4 - 5 | 583 | 121 | ||
| BC120829.1 | - | 4 - 5 | 513 | 40129752 | 121 | |
| BC120831.1 | - | 4 - 5 | 513 | 40129754 | 121 | |
| BC144835.1 | - | 4 - 5 | 513 | 121 | ||
More Information
Set Membership: |
|

Irak3
Genomic Map