Assay Details
Gene Symbol: Pag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195031.1 | NP_001181960.1 | 3 - 4 | 600 | 62 | |
NM_053182.5 | NP_444412.2 | 2 - 3 | 541 | 62 | ||
GenBank mRNA | AK040230.1 | - | 3 - 4 | 550 | 62 | |
AK089329.1 | - | 2 - 3 | 493 | 62 | ||
AK156000.1 | - | 3 - 4 | 552 | 62 | ||
AK156986.1 | - | 3 - 4 | 525 | 62 | ||
AK172106.1 | - | 3 - 4 | 523 | 62 | ||
BC145328.1 | - | 3 - 4 | 424 | 9053884 | 62 | |
BC145761.1 | - | 3 - 4 | 424 | 40131064 | 62 | |
BC145763.1 | - | 3 - 4 | 424 | 40131066 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK040230.1, AK089329.1, AK156000.1, AK172106.1, BC145328.1, BC145761.1, BC145763.1, NM_001195031.1, NM_053182.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
