Assay Details
Gene Symbol: Rp2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290643.1 | NP_001277572.1 | 2 - 3 | 1008 | 124 | |
NM_001290644.1 | NP_001277573.1 | 3 - 4 | 1167 | 124 | ||
NM_133669.5 | NP_598430.1 | 2 - 3 | 1008 | 124 | ||
NR_110966.1 | 2 - 3 | 1008 | 124 | |||
NR_110967.1 | 2 - 3 | 1008 | 124 | |||
GenBank mRNA | AJ303371.1 | - | 2 - 3 | 834 | 124 | |
AK017422.1 | - | 2 - 3 | 122 | |||
AK033954.1 | - | 3 - 4 | 1090 | 124 | ||
AK044002.1 | - | 2 - 3 | 915 | 124 | ||
AK050418.1 | - | 2 - 3 | 882 | 124 | ||
AK076907.1 | - | 2 - 3 | 870 | 124 | ||
AK132713.1 | - | 2 - 3 | 1008 | 124 | ||
AK162309.1 | - | 2 - 3 | 926 | 124 | ||
BC030300.1 | - | 1 - 2 | 610 | 124 | ||
BC049698.1 | - | 2 - 3 | 886 | 6706696 | 124 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -



Gene Ontology Categories:
MicroArray Probes:
