Assay Details
| Gene Symbol: Myrip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144557.5 | NP_653140.1 | 13 - 14 | 2413 | 126 | |
| GenBank mRNA | AB083782.1 | - | 12 - 13 | 2252 | 126 | |
| AF396688.1 | - | 12 - 13 | 2275 | 126 | ||
| AK038988.1 | - | 13 - 14 | 2367 | 126 | ||
| AK083225.1 | - | 13 - 14 | 2414 | 126 | ||
| AK148358.1 | - | 13 - 14 | 2530 | 126 | ||
| AY099470.1 | - | 12 - 13 | 2288 | 126 | ||
| BC129851.1 | - | 12 - 13 | 2297 | 40109680 | 126 | |
| BC129852.1 | - | 12 - 13 | 2297 | 40109682 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Myrip
Genomic Map