Assay Details
| Gene Symbol: Lpcat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145376.5 | NP_663351.3 | 2 - 3 | 283 | 58 | |
| GenBank mRNA | AB244717.2 | - | 2 - 3 | 283 | 58 | |
| AK081865.1 | - | 2 - 3 | 441 | 58 | ||
| AK132379.1 | - | 2 - 3 | 417 | 58 | ||
| AK155286.1 | - | 2 - 3 | 447 | 58 | ||
| AK169190.1 | - | 2 - 3 | 416 | 58 | ||
| AK170723.1 | - | 2 - 3 | 436 | 58 | ||
| AK171582.1 | - | 2 - 3 | 436 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lpcat1
Genomic Map