Assay Details
| Gene Symbol: Vac14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146216.2 | NP_666328.2 | 3 - 4 | 678 | 77 | |
| GenBank mRNA | AK031952.1 | - | 3 - 4 | 713 | 77 | |
| AK043319.1 | - | 3 - 4 | 710 | 77 | ||
| AK046826.1 | - | 3 - 4 | 650 | 77 | ||
| AK140191.1 | - | 3 - 4 | 705 | 77 | ||
| AK171577.1 | - | 3 - 4 | 718 | 77 | ||
| AK171883.1 | - | 3 - 4 | 705 | 77 | ||
| AK205932.1 | - | 1 - 2 | 38 | 77 | ||
| AK219487.1 | - | 1 - 2 | 38 | 77 | ||
| BC052199.1 | - | 3 - 4 | 678 | 6512678 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Vac14
Genomic Map