Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900007367.mm.1 |
NM_146224.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007367.mm.1 |
TC0900000959.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007370.mm.1 |
NM_146224.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007370.mm.1 |
TC0900000959.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007366.mm.1 |
NM_146224.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007366.mm.1 |
TC0900000959.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900007369.mm.1 |
NM_146224.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900007369.mm.1 |
TC0900000959.mm.1 |
MTA TC |