Assay Details
| Gene Symbol: Zbtb24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153398.3 | NP_700447.2 | 4 - 5 | 1408 | 100 | |
| NR_102360.1 | 4 - 5 | 1403 | 100 | |||
| GenBank mRNA | AK054097.1 | - | 4 - 5 | 1364 | 100 | |
| AK146986.1 | - | 4 - 5 | 1384 | 100 | ||
| AK154467.1 | - | 4 - 5 | 1379 | 100 | ||
| AY210402.1 | - | 4 - 5 | 1382 | 100 | ||
| BC050933.1 | - | 4 - 5 | 1370 | 6811382 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb24
Genomic Map